missing translation for 'onlineSavingsMsg'
Learn More

Thymol, 98+%

CAS: 89-83-8 | C10H14O | 150.221 g/mol

Supplier:  Thermo Scientific Chemicals A1456330

 View more versions of this product

Catalog No. AAA1456330

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Thymol is used as a preservative in halothane. It acts as an anesthetic, antiseptic in mouth wash, stabilizer in pharmaceutical preparations. It inhibits the growth and lactate production as well as reduces the uptake of cellular glucose within the bacteria. It is an active ingredient in toothpastes like euthymol. It is involved to control varroa mites and prevent fermentation and the growth of mold in bee colonies.

Solubility
Slightly soluble in water. Soluble in chloroform, alcohol and ether.

Notes
Incompatible with strong oxidizing agents and bases.
Specifications

Specifications

Thymol
231°C to 233°C
C10H14O
UN2430
14,9399
Slightly soluble in water. Soluble in chloroform,alcohol and ether.
CC1=CC(=C(C=C1)C(C)C)O
150.221
CHEBI:27607
≥98%
5 to 7
102°C (215°F)
49°C to 52°C
89-83-8
MFCD00002309
1907135
2-Isopropyl-5-methylphenol
MGSRCZKZVOBKFT-UHFFFAOYSA-N
5-methyl-2-propan-2-ylphenol
6989
150.22
0.97
250 g

Chemical Identifiers

89-83-8
150.221
MGSRCZKZVOBKFT-UHFFFAOYSA-N
6989
5-methyl-2-propan-2-ylphenol
C10H14O
MFCD00002309
2-Isopropyl-5-methylphenol
CHEBI:27607
CC1=CC(=C(C=C1)C(C)C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335-H500

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.

EINECSNumber : 201-944-8

RTECSNumber : XP2275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only