missing translation for 'onlineSavingsMsg'
Learn More

Thymol, 98+%

Catalog No. p-7021993
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1456322 100 g
AAA1456330 250 g
AAA1456336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1456322 Supplier Thermo Scientific Chemicals Supplier No. A1456322

CAS: 89-83-8 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Thymol is used as a preservative in halothane. It acts as an anesthetic, antiseptic in mouth wash, stabilizer in pharmaceutical preparations. It inhibits the growth and lactate production as well as reduces the uptake of cellular glucose within the bacteria. It is an active ingredient in toothpastes like euthymol. It is involved to control varroa mites and prevent fermentation and the growth of mold in bee colonies.

Solubility
Slightly soluble in water. Soluble in chloroform, alcohol and ether.

Notes
Incompatible with strong oxidizing agents and bases.

Chemical Identifiers

CAS 89-83-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002309
InChI Key MGSRCZKZVOBKFT-UHFFFAOYSA-N
Synonym 2-Isopropyl-5-methylphenol
PubChem CID 6989
ChEBI CHEBI:27607
IUPAC Name 5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1)C(C)C)O

Specifications

Melting Point 49°C to 52°C
Boiling Point 231°C to 233°C
UN Number UN2430
Beilstein 1907135
Merck Index 14,9399
Solubility Information Slightly soluble in water. Soluble in chloroform,alcohol and ether.
Formula Weight 150.22
Percent Purity ≥98%
Density 0.97
pH 5 to 7
Quantity 100 g
Flash Point 102°C (215°F)
Chemical Name or Material Thymol
Show More Show Less
Hazard Category H302-H314-H335-H500
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 201-944-8
RTECSNumber XP2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only