missing translation for 'onlineSavingsMsg'
Learn More

Thymine 98.0+%, TCI America™

Catalog No. p-7136159
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02345G 5 g
T023425G 25 g
T0234100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T02345G Supplier TCI America Supplier No. T02345G
Only null left

Chemical Identifiers

CAS 65-71-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00006026
InChI Key RWQNBRDOKXIBIV-UHFFFAOYSA-N
Synonym thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 1135
ChEBI CHEBI:17821
IUPAC Name 5-methyl-1H-pyrimidine-2,4-dione
SMILES CC1=CNC(=O)NC1=O

Specifications

Color White
Quantity 5 g
Formula Weight 126.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thymine
RTECSNumber XP2100000
TSCA Yes