missing translation for 'onlineSavingsMsg'
Learn More

Thymine, 99%

Catalog No. AC157850250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157850250 25 g Plastic bottle
AC157850050 5 g Glass Bottle
AC157851000 100 g Plastic bottle
AC157855000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC157850250 Supplier Thermo Scientific Chemicals Supplier No. 157850250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 65-71-4 | C5H6N2O2 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65-71-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006026
InChI Key RWQNBRDOKXIBIV-UHFFFAOYSA-N
Synonym thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 1135
ChEBI CHEBI:17821
IUPAC Name 5-methyl-1H-pyrimidine-2,4-dione
SMILES CC1=CNC(=O)NC1=O

Specifications

Melting Point 316.0°C to 317.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 24,353
Merck Index 15,9553
Solubility Information Solubility in water: slightly soluble (4g/L 25°C). Other solubilities: soluble in hot water, readily soluble in alkalies, sparingly soluble in ether, practically insoluble in alcohol
Formula Weight 126.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Thymine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-616-1

RUO – Research Use Only