missing translation for 'onlineSavingsMsg'
Learn More

Thymidine 5'-Monophosphate Disodium Salt Hydrate 98.0+%, TCI America™

Catalog No. T0845100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0845100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0845100MG Supplier TCI America Supplier No. T0845100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33430-62-5
Molecular Formula C10H13N2Na2O8P
Molecular Weight (g/mol) 366.17
MDL Number MFCD00023797
InChI Key AGSQMPPRYZYDFV-ZJWYQBPBSA-L
Synonym tmp disodium
PubChem CID 131674985
IUPAC Name disodium [(2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)NC1=O

Specifications

Color White
Quantity 100 mg
Formula Weight 366.17
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Thymidine 5′-Monophosphate Disodium Salt Hydrate
TSCA No
Recommended Storage Refrigerator