accessibility menu, dialog, popup

UserName

CAS RN 33430-62-5

IUPAC Name: disodium [(2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl phosphate
Synonyms: disodium thymidine 5'-monophosphate
Molecular Weight (g/mol): 366.17
Molecular Formula: C10H13N2Na2O8P
InChi Key: AGSQMPPRYZYDFV-ZJWYQBPBSA-L
SMILES: [Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)NC1=O
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)