missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymidine-5'-monophosphate disodium salt

Catalog No. p-7041927
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60747MC 100 mg
AAJ60747MD 250 mg
AAJ6074703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60747MC Supplier Thermo Scientific Chemicals Supplier No. J60747MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33430-62-5 | C10H13N2Na2O8P | 366.17 g/mol

Thymidine-5'-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymidine-5′-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.

Solubility
Soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 33430-62-5
Molecular Formula C10H13N2Na2O8P
Molecular Weight (g/mol) 366.17
MDL Number MFCD00023797
InChI Key AGSQMPPRYZYDFV-ZJWYQBPBSA-L
Synonym tmp disodium
PubChem CID 131674985
IUPAC Name disodium;[(2R,3S,5R)-3-hydroxy-5-(5-methyl-4-oxido-2-oxopyrimidin-1-yl)oxolan-2-yl]methyl hydrogen phosphate;hydrate
SMILES [Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)NC1=O

Specifications

Color White
Quantity 100 mg
Beilstein 4086927
Solubility Information Soluble in water.
Formula Weight 366.18
Physical Form Powder
Chemical Name or Material Thymidine-5'-monophosphate disodium salt
EINECSNumber 251-518-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only