missing translation for 'onlineSavingsMsg'
Learn More

Thulium, plasma standard solution, Specpure™ Tm 1000μg/mL

$188.89 - $188.89

Identifiants chimiques

MDL Number MFCD00011281
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA13824AE
Afficher les documents
Thermo Scientific Chemicals
013824AE
100 mL
chaque for $188.89
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

MFCD00011281

Spécifications

Liquid
MFCD00011281
100 mL
Thulium plasma standard solution, Tm 1000μg/mL
Tm2O3 in 5% HNO3
UN3264
Specpure™
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-714-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only