missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) acetate hydrate, REacton™, 99.9% (REO)

Catalog No. p-7049518
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1458209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1458209 Supplier Thermo Scientific Chemicals Supplier No. 01458209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 314041-04-8 | C6H9O6Tm | 346.07 g/mol

Thulium Acetate is the important dopant for silica-based fiber amplifiers. It can also be used in portable X-ray devices that has been bombarded in a nuclear reactor as a radiation source. Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. The other main application is used for optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 314041-04-8
Molecular Formula C6H9O6Tm
Molecular Weight (g/mol) 346.07
MDL Number MFCD00013046
InChI Key SNMVVAHJCCXTQR-UHFFFAOYSA-K
Synonym acmc-1afih, thulium acetate-water 1/3/1, thulium 3+ ion triacetate hydrate
IUPAC Name thulium(3+) triacetate
SMILES [Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Solubility Information Moderately soluble in water,strong mineral acids.
Formula Weight 346.07 (Anhydrous)
Percent Purity 99.9%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Thulium(III) acetate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-326-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only