accessibility menu, dialog, popup

UserName

CAS RN 314041-04-8

IUPAC Name: thulium(3+) triacetate
Synonyms: thulium(3+) triacetate
Molecular Weight (g/mol): 346.07
Molecular Formula: C6H9O6Tm
InChi Key: SNMVVAHJCCXTQR-UHFFFAOYSA-K
SMILES: [Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results