missing translation for 'onlineSavingsMsg'
Learn More

Thujone (alpha- and beta- mixture) 70.0+%, TCI America™

Catalog No. T09895ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09895ML 5 mL
T098925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09895ML Supplier TCI America Supplier No. T09895ML

Chemical Identifiers

CAS 1125-12-8
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00135597
InChI Key USMNOWBWPHYOEA-UHFFFAOYSA-N
Synonym 1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one
PubChem CID 11027
ChEBI CHEBI:50040
IUPAC Name 4-methyl-1-propan-2-ylbicyclo[3.1.0]hexan-3-one
SMILES CC1C2CC2(CC1=O)C(C)C

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 152.24
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Thujone (alpha- and beta- mixture)