accessibility menu, dialog, popup

UserName

H3CCH3CH3O

CAS RN 1125-12-8

IUPAC Name: 4-methyl-1-(propan-2-yl)bicyclo[3.1.0]hexan-3-one
Synonyms: 3-isothujone (1α,4α,5α)-β-thujone β-thujone, (1α,4α,5α)- 1s-(1α,4β,5α)-β-thujone chrysanthone tanacetone 3-thujanone (8CI) thujan-3-one 1S-(1α,4β,5α)-β-thujone β-thujone, (1α,4α,5α)-isomer
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: USMNOWBWPHYOEA-UHFFFAOYNA-N
SMILES: CC(C)C12CC1C(C)C(=O)C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results