missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THSD7A Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5149798

Catalog No. PA5149798

Only null left
Explore available promotions

Description

Description

THSD7A Polyclonal Antibody for Western Blot

The protein encoded by this gene is found almost exclusively in endothelial cells from placenta and umbilical cord. The encoded protein appears to interact with alpha(V)beta(3) integrin and paxillin to inhibit endothelial cell migration and tube formation. This protein may be involved in cytoskeletal organization. Variations in this gene may be associated with low bone mineral density in osteoporosis. [provided by RefSeq, Aug 2010].
Specifications

Specifications

THSD7A
Polyclonal
Unconjugated
THSD7A
KIAA0960; thrombospondin type 1 domain containing 7A; thrombospondin type-1 domain-containing protein 7A; Thrombospondin type-1 domain-containing protein 7A, soluble form; thrombospondin, type I, domain containing 7A; THSD7A
Rabbit
Protein A/G
RUO
221981, 330267, 500032
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q69ZU6, Q9UPZ6
THSD7A
Recombinant Thrombospondin Type I Domain Containing Protein 7A (THSD7A) (Thr42-Tyr242).
200 μL
Primary
Human, Mouse, Rat
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions