missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THRSP Polyclonal Antibody

Description
THRSP Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is similar to the gene product of S14, a rat gene whose expression is limited to liver and adipose tissue and is controlled by nutritional and hormonal factors. This gene has been shown to be expressed in liver and adipocytes, particularly in lipomatous modules. It is also found to be expressed in lipogenic breast cancers, which suggests a role in controlling tumor lipid metabolism.
Specifications
Specifications
| Antigen | THRSP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | THRSP |
| Gene Accession No. | Q62264, Q92748 |
| Gene Alias | lipogenic protein 1; Lipogenic protein S14; Lpgp; LPGP1; S14; S14MGC21659; Spot 14 protein; SPOT14; SPOT14 homolog; THRP; Thrsp; thyroid hormone responsive; thyroid hormone responsive (SPOT14 homolog, rat); thyroid hormone responsive SPOT14; thyroid hormone responsive SPOT14 homolog; thyroid hormone-inducible hepatic protein |
| Gene Symbols | THRSP |
| Show More |