missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thrombospondin 1 Monoclonal Antibody (C6.7)
GREENER_CHOICE

Catalog No. PIMA513390
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513390 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513390 Supplier Invitrogen™ Supplier No. MA513390
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MA5-13390 targets Thrombospondin in FACS, IF, IM, IP, and WB applications and shows reactivity with Bovine, Canine, Equine, Human, Ovine, Porcine, and Rat samples. The MA5-13390 immunogen is ca2+-replete, native purified human TSP from supernatant of thrombin-activated platelets.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Specifications

Antigen Thrombospondin 1
Applications Flow Cytometry, Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C6.7
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene THBS1
Gene Accession No. P07996, Q28178
Gene Alias glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1
Gene Symbols THBS1
Host Species Mouse
Immunogen Ca2+-replete, native purified human TSP from supernatant of thrombin-activated platelets.
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100057478, 101106433, 281530, 445442, 487486, 492313, 7057
Target Species Bovine, Canine, Horse, Human, Sheep, Pig, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less