missing translation for 'onlineSavingsMsg'
Learn More

Thrombopoietin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

$337.00 - $432.00

Specifications

Antigen Thrombopoietin
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
500P49GBT25 DFS Item
View Documents
PeproTech
500P49GBT25UG
25 μg
Each for $337.00
Only null left
 
500P49GBT50 DFS Item
View Documents
PeproTech
500P49GBT50UG
50 μg
Each for $432.00
Only null left
 
50-271-8478
View Documents
PeproTech
500P49GBT500UG
500 μg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

AA Sequence of recombinant protein: SPAPPACDLR VLSKLLRDSH VLHSRLSQCP EVHPLPTPVL LPAVDFSLGE WKTQMEETKA QDILGAVTLL LEGVMAARGQ LGPTCLSSLL GQLSGQVRLL LGALQSLLGT QLPPQGRTTA HKDPNAIFLS FQHLLRGKVR FLMLVGGSTL CVRRAPPTTA VPSRTSLVLT LNEL. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human TPO. Anti-Human TPO-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hTPO by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human TPO (500-P49G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hTPO. Western Blot: To detect hTPO by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hTPO is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P49GBT-1MG will be provided as 2 x 500 μg

Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, μgtimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
Specifications

Specifications

Thrombopoietin
ELISA, Western Blot
Biotin
Goat
Human
P40225
7066
E.coli-derived, 18.6 kDa Recombinant Human TPO.
Antigen affinity chromatography
THPO
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
C-mpl ligand; Megakaryocyte colony-stimulating factor; megakaryocyte growth and development factor; megakaryocyte stimulating factor; Mgdf; MKCSF; ML; MPL ligand; Mpllg; MSA; Myeloproliferative leukemia virus oncogene ligand; prepro-thrombopoietin; TDH2A; THCYT1; THPO; Thrombopoietin; thyroid microsomal antigen; thyroid peroxidase; thyroperoxidase; Tpo; TPX
THPO
Primary
-20°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'