missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Threonine (L), USP
Description
Threonine (L), USP comes as white odorless crystals. It is freely soluble in water, and insoluble in absolute alcohol.
Alternative name(s): L-Threonine, L-Thr
Specifications
Specifications
| Chemical Name or Material | Threonine (L), USP |
| Packaging | Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck |
| CAS | 72-19-5 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 119.1191 |
| Quantity | 25 g |
| Percent Purity | 98.5 to 101.5 % |
| Grade | USP |