missing translation for 'onlineSavingsMsg'
Learn More

Medisca Threonine (L), USP

Catalog No. 14185131
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185131 1 kg
14185431 100 g
14185451 25 g
14185169 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14185131 Supplier Medisca Supplier No. 030809

Threonine (L), USP comes as white odorless crystals. It is freely soluble in water, and insoluble in absolute alcohol.

Threonine (L), USP comes as white odorless crystals. It is freely soluble in water, and insoluble in absolute alcohol.

Alternative name(s): L-Threonine, L-Thr

Specifications

Chemical Name or Material Threonine (L), USP
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 72-19-5
Type Drug Standard
Molecular Weight (g/mol) 119.1191
Quantity 1 kg
Percent Purity 98.5 to 101.5 %
Grade USP