missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRAP3/TRAP150 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and X. laevis
THRAP3 is a 955 amino acid novel nuclear receptor and a member of the Thyroid hormone receptor associated proteins with a LISH domain and no obvious known sequence motifs. THRAP3 is a 150 kDa subunit of the large multiprotein complex TRAP and is important in the maintece of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It is also known to play an important role in transcriptional coactivation. It is known to interact with DKC1, nop5/nap65 and fibrillarin. From the Northern blot analysis of several human tissues it was found that TRAP150 is ubiquitously expressed.
Specifications
Specifications
| Antigen | THRAP3/TRAP150 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | THRAP3 |
| Gene Accession No. | Q9Y2W1 |
| Gene Alias | TRAP150, thyroid hormone receptor-associated protein 3, thyroid hormone receptor-associated protein complex 150 kDa component, thyroid hormone receptor-associated protein, 150 kDa subunit |
| Gene Symbols | THRAP3 |
| Show More |