missing translation for 'onlineSavingsMsg'
Learn More

Thioxanthone 98.0+%, TCI America™

Catalog No. t235125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
T235125G 25 g
1 options

Catalog No. T235125G

Supplier: TCI America T235125G

Only null left

Chemical Identifiers

CAS 492-22-8
Molecular Formula C13H8OS
Molecular Weight (g/mol) 212.27
MDL Number MFCD00005066
InChI Key YRHRIQCWCFGUEQ-UHFFFAOYSA-N
Synonym thioxanthone, 9h-thioxanthen-9-one, thiaxanthone, thioxanthenone, thiaxanthenone, thioxanthene-9-one, thiaxanthon, 9-thioxanthone, 10-thioxanthone, 9h-thioxanthene, 9-oxo
PubChem CID 10295
IUPAC Name 9H-thioxanthen-9-one
SMILES O=C1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 215°C
Color Yellow
Boiling Point 373°C
Quantity 25 g
Formula Weight 212.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thioxanthone
EINECSNumber (5)-3672
TSCA Yes