accessibility menu, dialog, popup

UserName

CAS RN 492-22-8

IUPAC Name: 9H-thioxanthen-9-one
Synonyms: thioxanthone
Molecular Weight (g/mol): 212.27
Molecular Formula: C13H8OS
InChi Key: YRHRIQCWCFGUEQ-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2SC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results