missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiourea Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 62-56-6 |
| Linear Formula | NH2 CSNH2 |
| MDL Number | MFCD00008067 |
| Melting Point | 170°C to 176°C |
| Synonym | Thiourea; Sulfourea; Thiocarbamide |
| Type | Certified Reference Material |
| Quantity | 200 mg |
| Formula Weight | 76.12 |
| Grade | Certified Reference Material |