missing translation for 'onlineSavingsMsg'
Learn More

Thiourea 99.0+%, TCI America™

Catalog No. T28355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28355G 5 g
T283525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28355G Supplier TCI America Supplier No. T28355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62-56-6
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00008067
InChI Key UMGDCJDMYOKAJW-UHFFFAOYSA-N
Synonym thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
PubChem CID 2723790
ChEBI CHEBI:36946
IUPAC Name thiourea
SMILES NC(N)=S

Specifications

Melting Point 177°C
Color White
pH 5.0 to 7.0 (50g/L H2O 25°C)
Quantity 5 g
UN Number 2811
Formula Weight 76.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiourea [for Biochemical Research]
EINECSNumber (2)-1733
RTECSNumber YU2800000
TSCA Yes