missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiosemicarbazide 98.0+%, TCI America™
Supplier: TCI America T0221100G
Specifications
| Thiosemicarbazide | |
| 184°C | |
| CH5N3S | |
| 100 g | |
| thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine | |
| C(=S)(N)NN | |
| 91.132 | |
| 91.13 | |
| Crystalline Powder |
| 79-19-6 | |
| White-Yellow | |
| MFCD00007620 | |
| 2811 | |
| BRWIZMBXBAOCCF-UHFFFAOYSA-N | |
| aminothiourea | |
| 2723789 | |
| ≥98.0% (T) |
Chemical Identifiers
| 79-19-6 | |
| 91.132 | |
| BRWIZMBXBAOCCF-UHFFFAOYSA-N | |
| 2723789 | |
| C(=S)(N)NN |
| CH5N3S | |
| MFCD00007620 | |
| thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine | |
| aminothiourea |
Safety and Handling
EINECSNumber : (2)-1746
RTECSNumber : VT4200000
TSCA : Yes