missing translation for 'onlineSavingsMsg'
Learn More

Thiosemicarbazide 98.0+%, TCI America™

Catalog No. p-7136151
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T022125G 25 g
T0221100G 100 g
T0221500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T022125G Supplier TCI America Supplier No. T022125G
Only null left

Chemical Identifiers

CAS 79-19-6
Molecular Formula CH5N3S
Molecular Weight (g/mol) 91.132
MDL Number MFCD00007620
InChI Key BRWIZMBXBAOCCF-UHFFFAOYSA-N
Synonym thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine
PubChem CID 2723789
IUPAC Name aminothiourea
SMILES C(=S)(N)NN

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 91.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiosemicarbazide
EINECSNumber (2)-1746
RTECSNumber VT4200000
TSCA Yes