missing translation for 'onlineSavingsMsg'
Learn More

Thiopropionamide 98.0+%, TCI America™

Catalog No. T066910G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T066910G 10 g
T066925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T066910G Supplier TCI America Supplier No. T066910G

Chemical Identifiers

CAS 631-58-3
Molecular Formula C3H7NS
Molecular Weight (g/mol) 89.156
MDL Number MFCD00059864
InChI Key WPZSAUFQHYFIPG-UHFFFAOYSA-N
PubChem CID 2760628
IUPAC Name propanethioamide
SMILES CCC(=S)N

Specifications

Melting Point 40°C
Quantity 10 g
Formula Weight 89.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thiopropionamide
TSCA No
Recommended Storage Refrigerator