missing translation for 'onlineSavingsMsg'
Learn More

Thiopicolinamide 97.0+%, TCI America™

Catalog No. T26181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26181G 1 g
T26185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26181G Supplier TCI America Supplier No. T26181G

Chemical Identifiers

CAS 5346-38-3
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.19
MDL Number MFCD00087576
InChI Key HYKQYVSNFPWGKQ-UHFFFAOYSA-N
Synonym thiopicolinamide, 2-pyridinecarbothioamide, pyridine-2-thioamide, pyridine-2-carbothioic acid amide, picolinamide, thio, 2-pyridinethiocarboxamide, picolinothioamide, 2-thiopicolinamide, pyridinecarbothioamide, 2-thiocarbamoylpyridine
PubChem CID 1549499
IUPAC Name pyridine-2-carbothioamide
SMILES NC(=S)C1=CC=CC=N1

Specifications

Melting Point 138°C
Quantity 1 g
Formula Weight 138.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thiopicolinamide
RTECSNumber TJ4305500
TSCA No