missing translation for 'onlineSavingsMsg'
Learn More

Thiophenol, MilliporeSigma™ Supelco™

$149.35 - $509.80

Specifications

CAS 108-98-5
Vapor Pressure 1.4 mmHg (20°C)
Vapor Density 3.8 (vs. air)
Density 1.073 g/mL (at 25°C)
Flash Point 50°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019943
View Documents
Merck Emd Millipore
092865MLF
5 mL
Each for $149.35
Only null left
 
111018402
View Documents
Merck Emd Millipore
0928625MLF
25 mL
Each for $509.80
Only null left
 
Description

Description

Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications

Specifications

108-98-5
3.8 (vs. air)
50°C
MFCD00004826
169°C
Analytical Standard
Analytical Standard
1.4 mmHg (20°C)
1.073 g/mL (at 25°C)
C6H5 SH
-15°C
Benzenethiol; Phenyl mercaptan
110.18
Videos
SDS
Documents

Documents