missing translation for 'onlineSavingsMsg'
Learn More

Thiophene, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020665
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020665 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020665 Supplier Merck Emd Millipore Supplier No. 069141MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 110-02-1
Vapor Pressure 40 mmHg (12.5°C)
Vapor Density 2.9 (vs. air)
Density 1.051 g/mL (at 25°C)
Linear Formula C4H4S
MDL Number MFCD00005413
Melting Point -38°C
Boiling Point 84°C
Synonym Thiofuran; Thiole
Quantity 1 mL
Formula Weight 84.14
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less