Learn More
Thiophene-3-carboxylic acid, 99%
CAS: 88-13-1 | C5H4O2S | 128.145 g/mol
Supplier: Thermo Scientific Chemicals A1571214
Description
3-Thiophenecarboxylic acid has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a lading compound for the development of a clinic useful D-amino acid inhibitor and have the potential to sever as active site proves to elucidate the structure-function relationships of D-amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Thiophenecarboxylic acid has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a lading compound for the development of a clinic useful D-amino acid inhibitor and have the potential to sever as active site proves to elucidate the structure-function relationships of D-amino acids.
Solubility
Soluble in water at 20°C 4.3 g/L.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Thiophene-3-carboxylic acid | |
| 137°C to 141°C | |
| MFCD00005467 | |
| 1994 | |
| 3-thiophenecarboxylic acid, 3-thenoic acid, 3-thiophenezoic acid, beta-thiophenic acid, 3-thienylcarboxylic acid, 3-thiophene carboxylic acid, 3-carboxythiophene, beta-thiophenecarboxylic acid, 3-thiophenoic acid, unii-6v3012q6be | |
| YNVOMSDITJMNET-UHFFFAOYSA-N | |
| thiophene-3-carboxylic acid | |
| 6918 | |
| 99% |
| 88-13-1 | |
| C5H4O2S | |
| 25 g | |
| 14,9279 | |
| Soluble in water at 20°C 4.3g/L. | |
| C1=CSC=C1C(=O)O | |
| 128.145 | |
| 128.15 |
Chemical Identifiers
| 88-13-1 | |
| 128.145 | |
| YNVOMSDITJMNET-UHFFFAOYSA-N | |
| 6918 | |
| C1=CSC=C1C(=O)O |
| C5H4O2S | |
| MFCD00005467 | |
| 3-thiophenecarboxylic acid, 3-thenoic acid, 3-thiophenezoic acid, beta-thiophenic acid, 3-thienylcarboxylic acid, 3-thiophene carboxylic acid, 3-carboxythiophene, beta-thiophenecarboxylic acid, 3-thiophenoic acid, unii-6v3012q6be | |
| thiophene-3-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-802-5
RTECSNumber : XM8330300
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only