missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carbonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. AAH6142814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6142814 25 g
AAH6142806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6142814 Supplier Thermo Scientific Chemicals Supplier No. H6142814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41507-35-1 | Molecular Formula: C5H3ClOS | Molecular Weight: 146.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41507-35-1
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00130090
InChI Key QTWBEVAYYDZLQL-UHFFFAOYSA-N
Synonym 3-thiophenecarbonyl chloride, 3-thenoyl chloride, 3-thiophenecarboxylic acid chloride, 3-thiophenecarbonylchloride, pubchem5597, 3-chlorocarbonyl thiophene, 3-thiophene carbonyl chloride, thiophene-3-carboxylic acid chloride, thiophene-3-carbonyl chloride, 3-thiophenecarbonyl chloride 6ci, 9ci
PubChem CID 2776377
IUPAC Name thiophene-3-carbonyl chloride
SMILES ClC(=O)C1=CSC=C1

Specifications

Melting Point 51°C to 54°C
Flash Point 51°C (124°F)
Quantity 25 g
UN Number UN2921
Sensitivity Moisture sensitive
Formula Weight 146.59
Percent Purity 97%
Chemical Name or Material Thiophene-3-carbonyl chloride
Hazard Category H228-H314-H335
Hazard Statement GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, FLAMMABLE, N.O.S.
EINECSNumber 255-420-9
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only