CAS RN 41507-35-1
IUPAC Name:
thiophene-3-carbonyl chloride
Synonyms:
thiophene-3-carbonyl chloride
Molecular Weight (g/mol):
146.59
Molecular Formula:
C5H3ClOS
InChi Key:
QTWBEVAYYDZLQL-UHFFFAOYSA-N
SMILES:
ClC(=O)C1=CSC=C1
1
–
1
of
1
results
1
Promotions Available
Thiophene-3-carbonyl chloride, 97%, Thermo Scientific Chemicals
Thiophene-3-carbonyl chloride, 97%, Thermo Scientific Chemicals
CAS: 41507-35-1 Molecular Formula: C5H3ClOS Molecular Weight (g/mol): 146.59 MDL Number: MFCD00130090 InChI Key: QTWBEVAYYDZLQL-UHFFFAOYSA-N Synonym: 3-thiophenecarbonyl chloride,3-thenoyl chloride,3-thiophenecarboxylic acid chloride,3-thiophenecarbonylchloride,pubchem5597,3-chlorocarbonyl thiophene,3-thiophene carbonyl chloride,thiophene-3-carboxylic acid chloride,thiophene-3-carbonyl chloride,3-thiophenecarbonyl chloride 6ci, 9ci PubChem CID: 2776377 IUPAC Name: thiophene-3-carbonyl chloride SMILES: ClC(=O)C1=CSC=C1