missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-acetic Acid 98.0+%, TCI America™

Catalog No. p-7136635
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11525G 5 g
T115225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11525G Supplier TCI America Supplier No. T11525G
Only null left

Chemical Identifiers

CAS 6964-21-2
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.172
MDL Number MFCD00005473
InChI Key RCNOGGGBSSVMAS-UHFFFAOYSA-N
Synonym 3-thiopheneacetic acid, 3-thienylacetic acid, 2-thiophen-3-yl acetic acid, thiophene-3-acetic acid, thiophen-3-yl-acetic acid, 3-thiophenacetic acid, thien-3-ylacetic acid, 3-thiopheneethanoic acid, thiophen-3-ylacetic acid, 2-3-thienyl acetic acid
PubChem CID 23404
IUPAC Name 2-thiophen-3-ylacetic acid
SMILES C1=CSC=C1CC(=O)O

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 129°C
Quantity 5 g
UN Number 3261
Formula Weight 142.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiophene-3-acetic Acid
RTECSNumber XM7570000
TSCA No