missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-acetic acid hydrazide, 97%

CAS: 175276-94-5 | C6H8N2OS | 156.20 g/mol

$148.64 - $619.62

Chemical Identifiers

CAS 175276-94-5
Molecular Formula C6H8N2OS
Molecular Weight (g/mol) 156.20
MDL Number MFCD00052946
InChI Key YNMRDHMMFCGQBD-UHFFFAOYSA-N
Synonym 2-thiophen-3-yl acetohydrazide, thiophene-3-acetic acid hydrazide, 2-3-thienyl acetohydrazide, 2-thien-3-yl acetohydrazide, 3-thiopheneacetic acid,hydrazide, 3 thienyl-acetic acid hydrazide, 2-thien-3-ylacetohydrazide, acmc-20ajii, maybridge1_008350, 3-thienyl acetohydrazide
PubChem CID 468649
IUPAC Name 2-thiophen-3-ylacetohydrazide
SMILES NNC(=O)CC1=CSC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1207703
View Documents
Thermo Scientific Chemicals
L1207703
1 g
Each for $148.64
Only null left
 
AAL1207706
View Documents
Thermo Scientific Chemicals
L1207706
5 g
Each for $619.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

175276-94-5
156.20
YNMRDHMMFCGQBD-UHFFFAOYSA-N
468649
NNC(=O)CC1=CSC=C1
C6H8N2OS
MFCD00052946
2-thiophen-3-yl acetohydrazide, thiophene-3-acetic acid hydrazide, 2-3-thienyl acetohydrazide, 2-thien-3-yl acetohydrazide, 3-thiopheneacetic acid,hydrazide, 3 thienyl-acetic acid hydrazide, 2-thien-3-ylacetohydrazide, acmc-20ajii, maybridge1_008350, 3-thienyl acetohydrazide
2-thiophen-3-ylacetohydrazide

Specifications

175276-94-5
C6H8N2OS
1 g
2-thiophen-3-yl acetohydrazide, thiophene-3-acetic acid hydrazide, 2-3-thienyl acetohydrazide, 2-thien-3-yl acetohydrazide, 3-thiopheneacetic acid,hydrazide, 3 thienyl-acetic acid hydrazide, 2-thien-3-ylacetohydrazide, acmc-20ajii, maybridge1_008350, 3-thienyl acetohydrazide
NNC(=O)CC1=CSC=C1
156.20
156.21
Thiophene-3-acetic acid hydrazide
85°C to 86°C
MFCD00052946
120473
YNMRDHMMFCGQBD-UHFFFAOYSA-N
2-thiophen-3-ylacetohydrazide
468649
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only