accessibility menu, dialog, popup

UserName

SONHH2N

CAS RN 175276-94-5

IUPAC Name: 2-(thiophen-3-yl)acetohydrazide
Synonyms: 2-(thiophen-3-yl)acetohydrazide
Molecular Weight (g/mol): 156.2
Molecular Formula: C6H8N2OS
InChi Key: YNMRDHMMFCGQBD-UHFFFAOYSA-N
SMILES: NNC(=O)CC1=CSC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)