missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-sulfonyl chloride, 97%

Catalog No. AAA1555306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1555306 5 g
AAA1555314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1555306 Supplier Thermo Scientific Chemicals Supplier No. A1555306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16629-19-9 | C4H3ClO2S2 | 182.64 g/mol

2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, bases.

Chemical Identifiers

CAS 16629-19-9
Molecular Formula C4H3ClO2S2
Molecular Weight (g/mol) 182.64
MDL Number MFCD00005426
InChI Key VNNLHYZDXIBHKZ-UHFFFAOYSA-N
Synonym 2-thiophenesulfonyl chloride, 2-thienylsulfonyl chloride, thiophene-2-sulphonyl chloride, 2-thiophenesulfonylchloride, 2-thienylsulfonylchloride, 2-thiophene sulfonyl chloride, chloro-2-thienylsulfone, pubchem7737, acmc-209dtw, thiophenesulfonyl chloride
PubChem CID 85518
IUPAC Name thiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CS1

Specifications

Melting Point 29°C to 31°C
Boiling Point 80°C to 81°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.573
Quantity 5 g
UN Number UN3261
Beilstein 123536
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 182.65
Percent Purity 97%
Physical Form Crystals or Powder or Crystalline Powder or Chunks or Fused Solid
Chemical Name or Material Thiophene-2-sulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement

H314-H318

Causes severe skin burns and eye damage.

Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 240-677-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only