missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carbonyl chloride, 98%

CAS: 5271-67-0 | C5H3ClOS | 146.59 g/mol

Supplier:  Thermo Scientific Chemicals A1464122

 View more versions of this product

Catalog No. AAA1464122

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thiophene-2-carbonyl chloride
1.372
90°C (194°F)
1.59
100 g
110145
2-thiophenecarbonyl chloride, 2-thenoyl chloride, 2-thienylcarbonyl chloride, 2-thiophenecarbonylchloride, thiophenecarbonyl chloride, 2-thiophene carbonyl chloride, thiophene-2-carboxylic acid chloride, thenoyl chloride, thienoyl chloride, 2-thenoylchloride
ClC(=O)C1=CC=CS1
146.59
146.6
5271-67-0
105°C (30 mmHg)
C5H3ClOS
MFCD00005428
UN3265
Moisture sensitive
QIQITDHWZYEEPA-UHFFFAOYSA-N
thiophene-2-carbonyl chloride
78928
98%

Chemical Identifiers

5271-67-0
146.59
QIQITDHWZYEEPA-UHFFFAOYSA-N
78928
ClC(=O)C1=CC=CS1
C5H3ClOS
MFCD00005428
2-thiophenecarbonyl chloride, 2-thenoyl chloride, 2-thienylcarbonyl chloride, 2-thiophenecarbonylchloride, thiophenecarbonyl chloride, 2-thiophene carbonyl chloride, thiophene-2-carboxylic acid chloride, thenoyl chloride, thienoyl chloride, 2-thenoylchloride
thiophene-2-carbonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 226-092-4

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only