accessibility menu, dialog, popup

UserName

CAS RN 5271-67-0

IUPAC Name: thiophene-2-carbonyl chloride
Synonyms: thiophene-2-carbonyl chloride
Molecular Weight (g/mol): 146.59
Molecular Formula: C5H3ClOS
InChi Key: QIQITDHWZYEEPA-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CS1
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)