missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carbonyl chloride, 98%

Catalog No. AAA1464114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1464114 25 g
AAA1464122 100 g
AAA1464136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1464114 Supplier Thermo Scientific Chemicals Supplier No. A1464114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5271-67-0 | C5H3ClOS | 146.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5271-67-0
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00005428
InChI Key QIQITDHWZYEEPA-UHFFFAOYSA-N
Synonym 2-thiophenecarbonyl chloride, 2-thenoyl chloride, 2-thienylcarbonyl chloride, 2-thiophenecarbonylchloride, thiophenecarbonyl chloride, 2-thiophene carbonyl chloride, thiophene-2-carboxylic acid chloride, thenoyl chloride, thienoyl chloride, 2-thenoylchloride
PubChem CID 78928
IUPAC Name thiophene-2-carbonyl chloride
SMILES ClC(=O)C1=CC=CS1

Specifications

Density 1.372
Boiling Point 105°C (30 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.59
Quantity 25 g
UN Number UN3265
Beilstein 110145
Sensitivity Moisture sensitive
Formula Weight 146.6
Percent Purity 98%
Chemical Name or Material Thiophene-2-carbonyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 226-092-4
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only