missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carbonitrile, 98%

CAS: 1003-31-2 | C5H3NS | 109.146 g/mol

Supplier:  Thermo Scientific Chemicals A1193022

 View more versions of this product

Catalog No. AAA1193022


Only null left
Explore available promotions

Description

Description

Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2'-thienylpyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Thiophene-2-carbonitrile
1.171
81°C (178°F)
C5H3NS
MFCD00005416
UN3276
2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile
CUPOOAWTRIURFT-UHFFFAOYSA-N
thiophene-2-carbonitrile
66087
98%
1003-31-2
191°C to 193°C
Unpleasant
1.5645
100 g
107035
Slightly miscible with water.
C1=CSC(=C1)C#N
109.146
109.15

Chemical Identifiers

1003-31-2
109.146
CUPOOAWTRIURFT-UHFFFAOYSA-N
66087
C1=CSC(=C1)C#N
C5H3NS
MFCD00005416
2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile
thiophene-2-carbonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H312-H332-H315-H319
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H318-H335

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 213-706-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only