missing translation for 'onlineSavingsMsg'
Learn More

(±)-Thiopental Solution, 1.0 mg/mL in Methanol, For Drug Analysis, Analytical Standard, ≥98%, MilliporeSigma™ Supelco™

Catalog No. 111017186
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017186 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017186 Supplier Merck Emd Millipore Supplier No. T10221ML
Only null left

Specifications

Packaging Ampule
CAS 76-75-5
Concentration 1.0 mg/mL in methanol
MDL Number MFCD00057564
Industry Type Drug Analysis
Quantity 1 mL
Percent Purity ≥98%
Grade Analytical Standard