Learn More
Thermo Scientific Chemicals Thionin acetate, pure, high purity biological stain
CAS: 78338-22-4 | C14H13N3O2S | 287.337 g/mol
$137.63 - $458.94
Chemical Identifiers
| CAS | 78338-22-4 |
|---|---|
| Molecular Formula | C14H13N3O2S |
| Molecular Weight (g/mol) | 287.337 |
| MDL Number | MFCD00081194 |
| InChI Key | JOIRQYHDJINFGA-UHFFFAOYSA-N |
| Synonym | 3, 7-Diaminophenothiazin-5-ium acetate |
| PubChem CID | 2724414 |
| IUPAC Name | (7-aminophenothiazin-3-ylidene)azanium;acetate |
| SMILES | CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC229840050
|
Thermo Scientific Chemicals
229840050 |
5 g | Glass bottle |
Each for $137.63
|
|
||||
|
AC229840250
|
Thermo Scientific Chemicals
229840250 |
25 g | Glass bottle |
Each for $458.94
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 78338-22-4 | |
| 287.337 | |
| JOIRQYHDJINFGA-UHFFFAOYSA-N | |
| 2724414 | |
| CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2 |
| C14H13N3O2S | |
| MFCD00081194 | |
| 3, 7-Diaminophenothiazin-5-ium acetate | |
| (7-aminophenothiazin-3-ylidene)azanium;acetate |
Specifications
| 78338-22-4 | |
| C14H13N3O2S | |
| 27, 392 | |
| 3, 7-Diaminophenothiazin-5-ium acetate | |
| JOIRQYHDJINFGA-UHFFFAOYSA-N | |
| CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2 | |
| 287.337 | |
| 597 to 601nm (in water) | |
| Pure | |
| Complies | |
| 5 g | |
| Thionin acetate, Pure |
| 85% min. | |
| MFCD00081194 | |
| 14, 9346 | |
| Solubility in water: ca 2.5g/L (25°C). Other solubilities: practically insoluble in ethanol | |
| Authentic | |
| (7-aminophenothiazin-3-ylidene)azanium;acetate | |
| 2724414 | |
| 287.33 | |
| Glass bottle | |
| Green | |
| Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only