missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiomorpholinoacetic Acid 1',1'-Dioxide Monohydrate 98.0+%, TCI America™
$202.15 - $610.28
Chemical Identifiers
| CAS | 155480-08-3 |
|---|---|
| Molecular Formula | C6H11NO4S |
| Molecular Weight (g/mol) | 193.217 |
| MDL Number | MFCD00121261 |
| InChI Key | CEABBIBOUBCOPV-UHFFFAOYSA-N |
| Synonym | 4-(Carboxymethyl)thiomorpholine 1,1-Dioxide, (1,1-Dioxothiomorpholino)acetic Acid |
| PubChem CID | 2806236 |
| IUPAC Name | 2-(1,1-dioxo-1,4-thiazinan-4-yl)acetic acid |
| SMILES | C1CS(=O)(=O)CCN1CC(=O)O |
Chemical Identifiers
| 155480-08-3 | |
| 193.217 | |
| CEABBIBOUBCOPV-UHFFFAOYSA-N | |
| 2806236 | |
| C1CS(=O)(=O)CCN1CC(=O)O |
| C6H11NO4S | |
| MFCD00121261 | |
| 4-(Carboxymethyl)thiomorpholine 1,1-Dioxide, (1,1-Dioxothiomorpholino)acetic Acid | |
| 2-(1,1-dioxo-1,4-thiazinan-4-yl)acetic acid |
Specifications
| 155480-08-3 | |
| White | |
| MFCD00121261 | |
| 4-(Carboxymethyl)thiomorpholine 1,1-Dioxide, (1,1-Dioxothiomorpholino)acetic Acid | |
| C1CS(=O)(=O)CCN1CC(=O)O | |
| 193.217 | |
| 193.22 | |
| Crystalline Powder |
| 177°C | |
| C6H11NO4S | |
| 5 g | |
| CEABBIBOUBCOPV-UHFFFAOYSA-N | |
| 2-(1,1-dioxo-1,4-thiazinan-4-yl)acetic acid | |
| 2806236 | |
| ≥98.0% (GC,T) | |
| Thiomorpholinoacetic Acid 1′,1′-Dioxide Monohydrate |
Safety and Handling
TSCA : No