accessibility menu, dialog, popup

UserName

CAS RN 155480-08-3

SOONOHO

CAS RN 155480-08-3

IUPAC Name: 2-(1,1-dioxo-1λ⁶-thiomorpholin-4-yl)acetic acid
Synonyms: (1,1-dioxo-1λ⁶-thiomorpholin-4-yl)acetic acid
Molecular Weight (g/mol): 193.22
Molecular Formula: C6H11NO4S
InChi Key: CEABBIBOUBCOPV-UHFFFAOYSA-N
SMILES: OC(=O)CN1CCS(=O)(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results