missing translation for 'onlineSavingsMsg'
Learn More

Thiomalic Acid 98.0+%, TCI America™

Catalog No. M006425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M006425G 25 g
M0064100G 100 g
M0064500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M006425G Supplier TCI America Supplier No. M006425G

Chemical Identifiers

CAS 70-49-5
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.148
MDL Number MFCD00004860
InChI Key NJRXVEJTAYWCQJ-UHFFFAOYSA-N
Synonym mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid
PubChem CID 6268
ChEBI CHEBI:38705
IUPAC Name 2-sulfanylbutanedioic acid
SMILES C(C(C(=O)O)S)C(=O)O

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 25 g
Formula Weight 150.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiomalic Acid
EINECSNumber (2)-1451
RTECSNumber WM8225000
TSCA Yes