accessibility menu, dialog, popup

UserName

CAS RN 70-49-5

IUPAC Name: 2-sulfanylbutanedioic acid
Synonyms: mercaptosuccinic acid thiomalic acid usaf M-2 (1)-mercaptosuccinic acid
Molecular Weight (g/mol): 150.15
Molecular Formula: C4H6O4S
InChi Key: NJRXVEJTAYWCQJ-UHFFFAOYNA-N
SMILES: OC(=O)CC(S)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (6)