missing translation for 'onlineSavingsMsg'
Learn More

Thioisonicotinamide 97.0+%, TCI America™

Catalog No. T07645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07645G 5 g
T076425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07645G Supplier TCI America Supplier No. T07645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2196-13-6
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.188
MDL Number MFCD00006437
InChI Key KPIIGXWUNXGGCP-UHFFFAOYSA-N
Synonym thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio
PubChem CID 2723788
IUPAC Name pyridine-4-carbothioamide
SMILES C1=CN=CC=C1C(=S)N

Specifications

Melting Point 200°C
Quantity 5 g
Formula Weight 138.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thioisonicotinamide
RTECSNumber US4204000
TSCA Yes