missing translation for 'onlineSavingsMsg'
Learn More

Thioindigo, TCI America™

Supplier:  TCI America T148925G

Catalog No. T148925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Thioindigo
280°C
C16H8O2S2
25 g
JOUDBUYBGJYFFP-NXVVXOECSA-N
2-[(2Z)-3-oxo-2,3-dihydro-1-benzothiophen-2-ylidene]-2,3-dihydro-1-benzothiophen-3-one
5378456
Crystalline Powder
522-75-8
Purple-Red
MFCD00191692
Fluorescent Red Dye, Vat Red 41
O=C1\C(SC2=CC=CC=C12)=C1\SC2=CC=CC=C2C1=O
296.36
296.36

Chemical Identifiers

522-75-8
296.36
JOUDBUYBGJYFFP-NXVVXOECSA-N
5378456
O=C1\C(SC2=CC=CC=C12)=C1\SC2=CC=CC=C2C1=O
C16H8O2S2
MFCD00191692
Fluorescent Red Dye, Vat Red 41
2-[(2Z)-3-oxo-2,3-dihydro-1-benzothiophen-2-ylidene]-2,3-dihydro-1-benzothiophen-3-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-2214

RTECSNumber : DT4330000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions