missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiocarbohydrazide 98.0+%, TCI America™
$65.24 - $182.68
Chemical Identifiers
| CAS | 2231-57-4 |
|---|---|
| Molecular Formula | CH6N4S |
| Molecular Weight (g/mol) | 106.15 |
| MDL Number | MFCD00007616 |
| InChI Key | LJTFFORYSFGNCT-UHFFFAOYSA-N |
| Synonym | thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu |
| PubChem CID | 2724189 |
| ChEBI | CHEBI:9549 |
| IUPAC Name | 1,3-diaminothiourea |
| SMILES | NNC(=S)NN |
Chemical Identifiers
| 2231-57-4 | |
| 106.15 | |
| LJTFFORYSFGNCT-UHFFFAOYSA-N | |
| 2724189 | |
| 1,3-diaminothiourea |
| CH6N4S | |
| MFCD00007616 | |
| thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu | |
| CHEBI:9549 | |
| NNC(=S)NN |
Specifications
| 2231-57-4 | |
| White-Yellow | |
| MFCD00007616 | |
| 2811 | |
| LJTFFORYSFGNCT-UHFFFAOYSA-N | |
| 1,3-diaminothiourea | |
| 2724189 | |
| 106.15 | |
| Crystalline Powder |
| 170°C | |
| CH6N4S | |
| 5 g | |
| thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu | |
| NNC(=S)NN | |
| 106.15 | |
| CHEBI:9549 | |
| ≥98.0% (T) | |
| Thiocarbohydrazide |
Safety and Handling
EINECSNumber : (2)-1745
RTECSNumber : FF2975000
TSCA : Yes