missing translation for 'onlineSavingsMsg'
Learn More

Thiocarbohydrazide 98.0+%, TCI America™

Catalog No. T11365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11365G 5 g
T113625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11365G Supplier TCI America Supplier No. T11365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2231-57-4
Molecular Formula CH6N4S
Molecular Weight (g/mol) 106.15
MDL Number MFCD00007616
InChI Key LJTFFORYSFGNCT-UHFFFAOYSA-N
Synonym thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu
PubChem CID 2724189
ChEBI CHEBI:9549
IUPAC Name 1,3-diaminothiourea
SMILES NNC(=S)NN

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 106.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiocarbohydrazide
EINECSNumber (2)-1745
RTECSNumber FF2975000
TSCA Yes